1-Ethylcyclopentyl methacrylate 98+% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A538249-25G A538249-1G
77569-046EA 196 USD
77569-046 77884-338
1-Ethylcyclopentyl methacrylate 98+% stabilized
1-Ethylcyclopentyl methacrylate

Precaución: For research use only.

Formula: C₁₁H₁₈O₂
MDL Number: MFCD23703079
CAS Number: 266308-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR