Potassium selenocyanate 99+%

Supplier: Ambeed
Danger

A530488-1G A530488-5G A530488-25G A530488-100G
77569-020EA 11 USD
77569-020 77602-262 77602-260 77880-150
Potassium selenocyanate 99+%
Potassium selenocyanate

Precaución: For research use only.

Formula: CKNSe
MDL Number: MFCD00011366
CAS Number: 3425-46-5
UN: 3283
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR