Trifluorolactic acid 98%

Supplier: Ambeed
Danger

Synonyms: 3,3,3-Trifluoro-2-hydroxypropanoic acid

A525993-1G A525993-5G
77569-012EA 32.96 USD
77569-012 77617-242
Trifluorolactic acid 98%
Trifluorolactic acid

Precaución: For research use only.

Formula: C₃H₃F₃O₃
MW: 144.05 g/mol
MDL Number: MFCD00051802
CAS Number: 684-07-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR