tert-Butyl 4-hydrazinylpiperidine-1-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A521441-1G A521441-250MG
77569-008EA 83 USD
77569-008 77893-063
tert-Butyl 4-hydrazinylpiperidine-1-carboxylate hydrochloride 95%

Precaución: For research use only.

Formula: C₁₀H₂₂ClN₃O₂
MDL Number: MFCD20921814
CAS Number: 1258001-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR