Alloc-Gly-OH ≥98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with TBC

A521251-1G A521251-5G
77569-006EA 47.38 USD
77569-006 77572-650
Alloc-Gly-OH ≥98% stabilized
Alloc-Gly-OH

Precaución: For research use only.

Formula: C₆H₉NO₄
MW: 159.1 g/mol
MDL Number: MFCD02094411
CAS Number: 90711-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR