N,N'-Bis(acryloyl)cystamine 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-(Disulphanediylbis(ethane-2,1-diyl))diacrylamide

A516852-25G A516852-1G A516852-5G A516852-250MG
77569-000EA 169.95 USD
77569-000 77602-220 77602-222 77669-300
N,N'-Bis(acryloyl)cystamine 98%
N,N'-Bis(acryloyl)cystamine

Precaución: For research use only.

Formula: C₁₀H₁₆N₂O₂S₂
MW: 260.37 g/mol
MDL Number: MFCD00036225
CAS Number: 60984-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR