(2R)-1,1,1-Trifluoropropan-2-ol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-1,1,1-Trifluoropropan-2-ol
A511893-1G
A511893-5G
A511893-100MG
77568-996EA
99.91
USD
77568-996
77568-998
77613-610
(2R)-1,1,1-Trifluoropropan-2-ol 98%
(2R)-1,1,1-Trifluoropropan-2-ol
Precaución: For research use only.
Formula:
C₃H₅F₃O MW: 114.066 g/mol |
MDL Number:
MFCD06797330 CAS Number: 17628-73-8 UN: 1987 ADR: 3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...