1,1'-(Methylenebis(4,1-phenylene))bis(2-hydroxy-2-methylpropan-1-one) 98%

Supplier: Ambeed
Warning

A505331-25G A505331-100G
77568-984EA 20 USD
77568-984 77594-584
1,1'-(Methylenebis(4,1-phenylene))bis(2-hydroxy-2-methylpropan-1-one) 98%
1,1'-(Methylenebis(4,1-phenylene))bis(2-hydroxy-2-methylpropan-1-one)

Precaución: For research use only.

Formula: C₂₁H₂₄O₄
MDL Number: MFCD16621105
CAS Number: 474510-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR