Fmoc-L-Gla(OtBu)2-OH 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-γ-carboxy-Glu(OtBu)₂-OH, Fmoc-Gla(OtBu)₂-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-4-(tert-butoxycarbonyl)-5-oxopentanoic acid

A501568-1G A501568-100MG A501568-50MG
77568-974EA 1496 USD
77568-974 77880-086 77880-087
Fmoc-L-Gla(OtBu)2-OH 95%
Fmoc-L-Gla(OtBu)2-OH

Precaución: For research use only.

Formula: C₂₉H₃₅NO₈
MDL Number: MFCD00038770
CAS Number: 111662-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR