alpha-Tocopherol polyethylene glycol succinate α-Tocopherol ≥ 25%

Supplier: Ambeed
Warning

Synonyms: Tocofersolan, Vitamin E Polyethylene Glycol Succinate, α-Tocopherol polyethylene glycol succinate

A493138-25G A493138-100G A493138-5G A493138-1G A493138-500G
77568-966EA 53 USD
77568-966 77572-562 77572-564 77602-128 77880-076
alpha-Tocopherol polyethylene glycol succinate α-Tocopherol ≥ 25%
alpha-Tocopherol polyethylene glycol succinate

Precaución: For research use only.

Formula: (C₂H₄O)nC₃₃H₅₄O₅
CAS Number: 9002-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR