4-Phenylbenzylamine 98%

Supplier: Ambeed
Warning

A491837-1G A491837-250MG
77568-962EA 10 USD
77568-962 77749-004
4-Phenylbenzylamine 98%
4-Phenylbenzylamine

Precaución: For research use only.

Formula: C₁₃H₁₃N
MDL Number: MFCD01310838
CAS Number: 712-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR