4-(((tert-Butyldimethylsilyl)oxy)methyl)phenol 97%

Supplier: Ambeed
Warning

A461880-1G A461880-5G
77568-910EA 69.01 USD
77568-910 77568-912
4-(((tert-Butyldimethylsilyl)oxy)methyl)phenol 97%
4-(((tert-Butyldimethylsilyl)oxy)methyl)phenol

Precaución: For research use only.

Formula: C₁₃H₂₂O₂Si
MW: 238.398 g/mol
MDL Number: MFCD29042407
CAS Number: 126070-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR