4-Bromo-2-fluoropyrimidine 95%

Supplier: Ambeed
Warning

A459489-250MG A459489-1G
77568-906EA 708 USD
77568-906 77884-182
4-Bromo-2-fluoropyrimidine 95%
4-Bromo-2-fluoropyrimidine

Precaución: For research use only.

Formula: C₄H₂BrFN₂
MDL Number: MFCD14702762
CAS Number: 1209458-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR