2-Cyano-5-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

A455914-1G A455914-250MG
77568-896EA 91 USD
77568-896 77568-898
2-Cyano-5-fluorobenzoic acid 95%
2-Cyano-5-fluorobenzoic acid

Precaución: For research use only.

Formula: C₈H₄FNO₂
MDL Number: MFCD03094320
CAS Number: 518070-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR