2-Fluoro-6-methoxybenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)-1-fluoro-3-methoxybenzene

A452232-250MG A452232-1G
77568-890EA 24 USD
77568-890 77572-422
2-Fluoro-6-methoxybenzyl bromide 98%
2-Fluoro-6-methoxybenzyl bromide

Precaución: For research use only.

Formula: C₈H₈BrFO
MDL Number: MFCD08688579
CAS Number: 500912-17-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR