4-Bromopyrimidine hydrobromide 95%

Supplier: Ambeed
Warning

A449742-1G A449742-100MG A449742-250MG
77568-884EA 51.5 USD
77568-884 77594-402 77669-024
4-Bromopyrimidine hydrobromide 95%
4-Bromopyrimidine hydrobromide

Precaución: For research use only.

Formula: C₄H₄Br₂N₂
MW: 239.896 g/mol
MDL Number: MFCD12913811
CAS Number: 1187931-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR