Methyl 2-formyl-3,5-dimethoxybenzoate 97%

Supplier: Ambeed
Warning

A448759-5G A448759-1G
77568-880EA 65 USD
77568-880 77669-016
Methyl 2-formyl-3,5-dimethoxybenzoate 97%
Methyl 2-formyl-3,5-dimethoxybenzoate

Precaución: For research use only.

Formula: C₁₁H₁₂O₅
MDL Number: MFCD01321304
CAS Number: 52344-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR