5-Iodo-3,4-dimethylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A433853-1G A433853-250MG
77568-866EA 407.88 USD
77568-866 77616-872
5-Iodo-3,4-dimethylpyridin-2-amine 95%
5-Iodo-3,4-dimethylpyridin-2-amine

Precaución: For research use only.

Formula: C₇H₉IN₂
MW: 248.06 g/mol
MDL Number: MFCD30737791
CAS Number: 2168839-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR