Ethyldimethylsilane 98%

Supplier: Ambeed
Danger

A422850-5G A422850-1G A422850-25G
77568-850EA 67 USD
77568-850 77882-102 77882-103
Ethyldimethylsilane 98%
Ethyldimethylsilane

Precaución: For research use only.

Formula: C₄H₁₂Si
MW: 88.22 g/mol
MDL Number: MFCD00009016
CAS Number: 758-21-4
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR