Difloxacin hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride

A418662-1G A418662-25G A418662-5G A418662-100G
77568-844EA 17 USD
77568-844 77626-840 77626-842 77879-915
Difloxacin hydrochloride 98+%
Difloxacin hydrochloride

Precaución: For research use only.

Formula: C₂₁H₁₉F₂N₃O₃·HCl
MDL Number: MFCD03840489
CAS Number: 91296-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR