(Bicyclo[1.1.1]pent-1-yl)methanol 95%

Supplier: Ambeed
Danger

Synonyms: Bicyclo[1.1.1]pentan-1-ylmethanol, 1-(Hydroxymethyl)bicyclo[1.1.1]pentane

A412674-250MG A412674-25MG
77568-830EA 337.84 USD
77568-830 77572-280
(Bicyclo[1.1.1]pent-1-yl)methanol 95%
(Bicyclo[1.1.1]pent-1-yl)methanol

Precaución: For research use only.

Formula: C₆H₁₀O
MW: 98.143 g/mol
MDL Number: MFCD28040494
CAS Number: 22287-32-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR