Butyl stearate 97%

Supplier: Ambeed
Warning

Synonyms: Stearic acid butyl ester, Butyl octadecanoate

A410472-1000G A410472-100G A410472-500G
77568-824EA 24 USD
77568-824 77572-274 77626-826
Butyl stearate 97%
Butyl stearate

Precaución: For research use only.

Formula: C₂₂H₄₄O₂
MW: 340.59 g/mol
Boiling Pt: 220
Melting Pt: 21
Flash Pt: 190
MDL Number: MFCD00026669
CAS Number: 123-95-5
Merck Index: 13,01589

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR