tert-Butyl 4-(prop-2-yn-1-yl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-4-(2-propynyl)piperazine

A396531-250MG A396531-1G
77568-804EA 51 USD
77568-804 77572-218
tert-Butyl 4-(prop-2-yn-1-yl)piperazine-1-carboxylate 95%
tert-Butyl 4-(prop-2-yn-1-yl)piperazine-1-carboxylate

Precaución: For research use only.

Formula: C₁₂H₂₀N₂O₂
MW: 224.299 g/mol
MDL Number: MFCD04039721
CAS Number: 199538-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR