tert-Butyl (6-bromonaphthalen-2-yl)carbamate 98%

Supplier: Ambeed
Warning

A386984-25G A386984-1G A386984-5G
77568-794EA 245 USD
77568-794 77890-801 77890-802
tert-Butyl (6-bromonaphthalen-2-yl)carbamate 98%
tert-Butyl (6-bromonaphthalen-2-yl)carbamate

Precaución: For research use only.

Formula: C₁₅H₁₆BrNO₂
MDL Number: MFCD13191642
CAS Number: 869114-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR