7-Bromofuro[3,2-c]pyridine 98%

Supplier: Ambeed
Warning

A385938-1G A385938-250MG
77568-782EA 767.35 USD
77568-782 77572-150
7-Bromofuro[3,2-c]pyridine 98%
7-Bromofuro[3,2-c]pyridine

Precaución: For research use only.

Formula: C₇H₄BrNO
MW: 198.01 g/mol
MDL Number: MFCD18254548
CAS Number: 603300-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR