4-((R)-Hydroxy((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl)quinolin-6-ol 98%

Supplier: Ambeed
Warning

A385598-5G A385598-250MG A385598-1G
77568-778EA 244.11 USD
77568-778 77594-172 77684-110
4-((R)-Hydroxy((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl)quinolin-6-ol 98%
4-((R)-Hydroxy((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl)quinolin-6-ol

Precaución: For research use only.

Formula: C₁₉H₂₂N₂O₂
MW: 310.39 g/mol
MDL Number: MFCD09842542
CAS Number: 524-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR