4-Bromo-2-fluoro-5-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-5-fluoro-4-methyl-2-nitrobenzene

A369594-25G A369594-5G
77568-756EA 53 USD
77568-756 77572-104
4-Bromo-2-fluoro-5-nitrotoluene 98%
4-Bromo-2-fluoro-5-nitrotoluene

Precaución: For research use only.

Formula: C₇H₅BrFNO₂
MDL Number: MFCD11042845
CAS Number: 1345471-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR