2-Amino-4-bromo-5-methylthiazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-5-methylthiazol-2-amine

A364556-1G A364556-250MG
77568-746EA 135.96 USD
77568-746 77668-688
2-Amino-4-bromo-5-methylthiazole 98%
2-Amino-4-bromo-5-methylthiazole

Precaución: For research use only.

Formula: C₄H₅BrN₂S
MW: 193.0700 g/mol
MDL Number: MFCD00995876
CAS Number: 1209167-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR