Oxetan-3-ylhydrazine dihydrochloride 95%

Supplier: Ambeed
Warning

A363166-1G A363166-250MG
77568-742EA 404 USD
77568-742 77890-746
Oxetan-3-ylhydrazine dihydrochloride 95%
Oxetan-3-ylhydrazine dihydrochloride

Precaución: For research use only.

Formula: C₃H₁₀Cl₂N₂O
MDL Number: MFCD22380551
CAS Number: 1374652-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR