3-(Trifluoromethyl)bicyclo[1.1.1]pentan-1-amine hydrochloride 98%

Supplier: Ambeed
Warning

A358606-1G A358606-100MG
77568-732EA 554.14 USD
77568-732 77686-016
3-(Trifluoromethyl)bicyclo[1.1.1]pentan-1-amine hydrochloride 98%
3-(Trifluoromethyl)bicyclo[1.1.1]pentan-1-amine hydrochloride

Precaución: For research use only.

Formula: C₆H₉ClF₃N
MW: 187.59 g/mol
MDL Number: MFCD22056474
CAS Number: 262852-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR