(5-(Trifluoromethyl)-1H-pyrazol-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A351704-100MG A351704-1G A351704-250MG A351704-5G
77568-702EA 57 USD
77568-702 77613-004 77883-963 77883-964
(5-(Trifluoromethyl)-1H-pyrazol-3-yl)boronic acid 97%
(5-(Trifluoromethyl)-1H-pyrazol-3-yl)boronic acid

Precaución: For research use only.

Formula: C₄H₄BF₃N₂O₂
MDL Number: MFCD28009360
CAS Number: 1346270-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR