7-Aminoimidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: Imidazo[1,2-a]pyridin-7-amine

A351378-1G A351378-250MG
77568-698EA 170.98 USD
77568-698 77668-620
7-Aminoimidazo[1,2-a]pyridine 98%
7-Aminoimidazo[1,2-a]pyridine

Precaución: For research use only.

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD11052644
CAS Number: 421595-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR