Dimethyl methoxymalonate 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl Methoxymalonate, Methoxymalonic acid dimethyl ester

A328896-5G A328896-25G A328896-100G
77568-666EA 77 USD
77568-666 77571-946 77879-694
Dimethyl methoxymalonate 95%
Dimethyl methoxymalonate

Precaución: For research use only.

Formula: C₆H₁₀O₅
MW: 162.14 g/mol
MDL Number: MFCD00009847
CAS Number: 5018-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR