5-Fluoro-2-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoropyridin-2-ylboronic acid

A324964-1G A324964-5G A324964-250MG
77568-652EA 143.17 USD
77568-652 77571-924 77668-456
5-Fluoro-2-pyridineboronic acid 98%
5-Fluoro-2-pyridineboronic acid

Precaución: For research use only.

Formula: C₅H₅BFNO₂
MDL Number: MFCD08705712
CAS Number: 946002-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR