Ethyl-5-bromo-1,3,4-oxadiazole-2-carboxylate 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-bromo-1,3,4-oxadiazole-2-carboxylate

A317402-1G A317402-100MG A317402-250MG
77568-638EA 167.89 USD
77568-638 77612-834 77683-944
Ethyl-5-bromo-1,3,4-oxadiazole-2-carboxylate 98%, Ambeed.Inc
Ethyl-5-bromo-1,3,4-oxadiazole-2-carboxylate

Precaución: For research use only.

Formula: C₅H₅BrN₂O₃
MW: 221.02 g/mol
MDL Number: MFCD22415767
CAS Number: 916889-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR