(Z)-4-Hydroxytamoxifen 99%

Supplier: Ambeed
Warning

Synonyms: (Z)-4-(1-(4-(2-(Dimethylamino)ethoxy)phenyl)-2-phenylbut-1-en-1-yl)phenol, (Z)-4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, (Z)-4-OHT, trans-4-Hydroxytamoxifen

A309666-50MG A309666-10MG
77568-624EA 376.98 USD
77568-624 77748-602
(Z)-4-Hydroxytamoxifen 99%
(Z)-4-Hydroxytamoxifen

Precaución: For research use only.

Formula: C₂₆H₂₉NO₂
MW: 387.51 g/mol
MDL Number: MFCD00278780
CAS Number: 68047-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR