Benzyl 3-amino-4-oxopiperidine-1-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A307358-1G A307358-250MG A307358-5G A307358-100MG
77568-618EA 122 USD
77568-618 77872-424 77872-425 77872-423
Benzyl 3-amino-4-oxopiperidine-1-carboxylate hydrochloride 95%
Benzyl 3-amino-4-oxopiperidine-1-carboxylate hydrochloride

Precaución: For research use only.

Formula: C₁₃H₁₇ClN₂O₃
MDL Number: MFCD12756125
CAS Number: 1196145-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR