4,4,5,5-Tetramethyl-2-(phenylethynyl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A301442-1G A301442-5G
77568-610EA 86 USD
77568-610 77885-799
4,4,5,5-Tetramethyl-2-(phenylethynyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(phenylethynyl)-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₁₄H₁₇BO₂
MDL Number: MFCD09842761
CAS Number: 159087-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR