Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥98% (by GC)
Supplier: Ambeed
|
Danger
|
Synonyms:
Ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate
A294463-25G
A294463-1G
77568-606EA
67.98
USD
77568-606
77748-575
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥98% (by GC)
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate
Precaución: For research use only.
Formula:
C₆H₆ClF₃O₃ MW: 218.556 g/mol |
MDL Number:
MFCD00041540 CAS Number: 363-58-6 UN: 1760 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...