Diethyl cyanophosphonate 95%

Supplier: Ambeed
Danger

Synonyms: Diethyl phosphorocyanidate

A279381-25G A279381-5G A279381-1G
77568-558EA 260 USD
77568-558 77571-736 77879-523
Diethyl cyanophosphonate 95%
Diethyl cyanophosphonate

Precaución: For research use only.

Formula: C₅H₁₀NO₃P
MDL Number: MFCD00010256
CAS Number: 2942-58-7
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR