(3-((Trimethylsilyl)ethynyl)phenyl)methanol 95%

Supplier: Ambeed
Warning

A277495-1G A277495-250MG
77568-544EA 209.09 USD
77568-544 77748-533
(3-((Trimethylsilyl)ethynyl)phenyl)methanol 95%
(3-((Trimethylsilyl)ethynyl)phenyl)methanol

Precaución: For research use only.

Formula: C₁₂H₁₆OSi
MW: 204.3403 g/mol
MDL Number: MFCD30537097
CAS Number: 1007587-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR