4-Amino-3-fluorobenzylalcohol 95%

Supplier: Ambeed
Warning

Synonyms: (4-Amino-3-fluorophenyl)methanol

A265854-1G A265854-250MG
77568-526EA 110 USD
77568-526 77601-140
4-Amino-3-fluorobenzylalcohol 95%
4-Amino-3-fluorobenzylalkohol

Precaución: For research use only.

Formula: C₇H₈FNO
MDL Number: MFCD03094241
CAS Number: 146019-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR