3-Chloro-N-fmoc-L-phenylalanine 98%

Supplier: Ambeed
Warning

A258839-25G A258839-100G A258839-10G A258839-1G
77568-494EA 189 USD
77568-494 77601-084 77881-699 77881-700
3-Chloro-N-fmoc-L-phenylalanine 98%
3-Chloro-N-fmoc-L-phenylalanine

Precaución: For research use only.

Formula: C₂₄H₂₀ClNO₄
MDL Number: MFCD00672548
CAS Number: 198560-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR