4-Nitro-5-trifluoromethyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitro-5-(Trifluoromethyl)-1H-pyrazole

A258300-5G A258300-1G
77568-492EA 220.42 USD
77568-492 77685-736
4-Nitro-5-trifluoromethyl-1H-pyrazole 98%
4-Nitro-5-trifluoromethyl-1H-pyrazole

Precaución: For research use only.

Formula: C₄H₂F₃N₃O₂
MW: 181.073 g/mol
MDL Number: MFCD04971110
CAS Number: 1046462-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR