2,9-Diphenyl-1,10-phenanthroline 98%

Supplier: Ambeed
Warning

A254381-1G A254381-5G
77568-464EA 126 USD
77568-464 77746-409
2,9-Diphenyl-1,10-phenanthroline 98%
2,9-Diphenyl-1,10-phenanthroline

Precaución: For research use only.

Formula: C₂₄H₁₆N₂
MDL Number: MFCD00044208
CAS Number: 25677-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR