Methyl 2-([1,1'-biphenyl]-4-yl)acetate 97%

Supplier: Ambeed
Warning

A251607-5G A251607-1G
77568-442EA 52 USD
77568-442 77892-463
Methyl 2-([1,1'-biphenyl]-4-yl)acetate 97%

Precaución: For research use only.

Formula: C₁₅H₁₄O₂
MDL Number: MFCD02056549
CAS Number: 59793-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR