tert-Butyl 3-(chlorosulfonyl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Danger

A249665-1G A249665-250MG
77568-424EA 275 USD
77568-424 77885-654
tert-Butyl 3-(chlorosulfonyl)pyrrolidine-1-carboxylate 97%
tert-Butyl 3-(chlorosulfonyl)pyrrolidine-1-carboxylate

Precaución: For research use only.

Formula: C₉H₁₆ClNO₄S
MDL Number: MFCD09909937
CAS Number: 935845-20-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR