2-(6-((tert-Butoxycarbonyl)amino)-9H-purin-9-yl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N6-Boc-adenosin-9-yl acetic acid
A249550-1G
A249550-5G
77568-422EA
203.94
USD
77568-422
77685-704
2-(6-((tert-Butoxycarbonyl)amino)-9H-purin-9-yl)acetic acid 97%
2-(6-((tert-Butoxycarbonyl)amino)-9H-purin-9-yl)acetic acid
Precaución: For research use only.
Formula:
C₁₂H₁₅N₅O₄ MW: 293.27 g/mol |
MDL Number:
MFCD24465997 CAS Number: 186046-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...