Methyl 2-(2-bromothiazol-4-yl)acetate 97%

Supplier: Ambeed
Warning

A249341-5G A249341-1G A249341-100MG A249341-250MG
77568-414EA 356 USD
77568-414 77612-406 77890-411 77890-412
Methyl 2-(2-bromothiazol-4-yl)acetate 97%
Methyl 2-(2-bromothiazol-4-yl)acetate

Precaución: For research use only.

Formula: C₆H₆BrNO₂S
MDL Number: MFCD18413039
CAS Number: 1261589-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR