(2-Hydroxylpropyl)-γ-cyclodextrine 98%

Supplier: Ambeed
Warning

Synonyms: (2-Hydroxypropyl)-γ-Cyclodextrin, HGC

A245744-25G A245744-100G A245744-1G A245744-5G
77568-392EA 58.71 USD
77568-392 77612-366 77612-368 77612-370
(2-Hydroxylpropyl)-γ-cyclodextrine 98%
(2-Hydroxylpropyl)-γ-cyclodextrine

Precaución: For research use only.

Formula: C₇₂H₁₂₈O₄₈
MW: 1761.7600 g/mol
MDL Number: MFCD00148890
CAS Number: 128446-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR